Få mere at vide
Atropine EP Impurity A (Hyoscyamine Sulfate Impurity G) as Hydrochloride, Mikromol™
Beskrivelse
Atropine EP Impurity A (Hyoscyamine Sulfate Impurity G) as Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (1R,3r,5S)-8-Methyl-8-azabicyclo(3.2.1)oct-3-yl 2-Phenylpropenoate Hydrochloride (Apoatropine Hydrochloride) |
| CAS | 5978-81-4 |
| Analyt- eller komponentnavne | (1R,3r,5S)-8-Methyl-8-azabicyclo(3.2.1)oct-3-yl 2-Phenylpropenoate Hydrochloride (Apoatropine Hydrochloride) |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C17 H21 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 2-Phenylpropenoate Hydrochloride (Apoatropine Hydrochloride), Atropine Imp. A (EP), Hyoscyamine Sulfate Imp. G (EP), Hyoscyamine Imp. G (EP), Apoatropine Hydrochloride, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 2-Phenylprop-2-enoate Hydrochloride, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 2-Phenylpropenoate Hydrochloride, Hyoscyamine Sulphate Imp. G (EP) as Hydrochloride, Atropine Imp. A (EP) as Hydrochloride, Atropine Impurity A as Hydrochloride, Hyoscyamine Sulfate Impurity G, Hyoscyamine Sulfate Impurity G as Hydrochloride, Hyoscyamine Impurity G, Hyoscyamine Impurity G as Hydrochloride |
| InChI formel | InChI=1S/C17H21NO2.ClH/c1-12(13-6-4-3-5-7-13)17(19)20-16-10-14-8-9-15(11-16)18(14)2;/h3-7,14-16H,1,8-11H2,2H3;1H/t14-,15+,16+; |
| SMIL | Cl.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)C(=C)c3ccccc3 |
| IUPAC navn | [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-phenylprop-2-enoate;hydrochloride |
| Vis mere |