Få mere at vide
Atrazine-d5, TRC
Artikelnummer. 30345496
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30345496 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345496 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7946022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Atrazine-d5
CAS 163165-75-1
Renhedsgrad noter HPLC
Molekylær formel C8 2H5 H9 Cl N5
Mængde 2.5 mg
Synonym Atrazine D5 (ethyl D5), 2-Chloro-4-pentadeuteroethylamino-6-isopropylamine-1,3,5-triazine, Atrazine-d5
InChI formel InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)/i1D3,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC(C)C)n1
IUPAC navn 6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 220.71
Formel vægt 220.1252
Procent renhed >95