Få mere at vide
Atractylenolide II, MedChemExpress
Artikelnummer. 16488888
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16488888 10 mg
16454787 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16488888 Leverandør Medchem Express Leverandørnr. HYN0202
MedChemExpress Atractylenolide II is a sesquiterpene compound isolated from the dried rhizome of Atractylodes macrocephala (Baizhu in Chinese); anti-proliferative activity.
Atractylenolide II is a sesquiterpene compound isolated from the dried rhizome of Atractylodes macrocephala (Baizhu in Chinese)
Tekniske data
Kemisk navn eller materiale Atractylenolide II
CAS 73069-14-4
Farve White
Renhedsgrad noter Research
Molekylær formel C15H20O2
Mængde 10 mg
Synonym Asterolide
Opløselighedsinformation DMSO : 100 mg/mL (430.44 mM; Need ultrasonic)
SMIL O=C1C(C)=C(C[C@@]23[H])[C@@](C[C@@]3(C)CCCC2=C)([H])O1
Molekylvægt (g/mol) 232.32
Formel vægt 232.32
Procent renhed 99.91%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease