missing translation for 'onlineSavingsMsg'
Få mere at vide

ATP8 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

1082.11 DKK - 3059.75 DKK

Tekniske data

Antigen ATP8
Koncentration 1000 μg/mL
Applikationer Western Blot, ELISA
Klassifikation Polyclonal
Konjugeret Unconjugated
Se flere specifikationer

Produkter 2
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
30236758
missing translation for 'viewDocuments'
Proteintech
29398-1-AP-150UL
150 μL
3059.75 DKK
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
30235945
missing translation for 'viewDocuments'
Proteintech
29398-1-AP-20UL
20 μL
1082.11 DKK
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Description

Description

The ATP8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATP8. This antibody recognizes human antigen. The ATP8 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

ATP8
Western Blot, ELISA
Unconjugated
Rabbit
Human
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1000 μg/mL
Polyclonal
Liquid
RUO
4509
IgG
Antigen affinity purification
Polyclonal Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.