missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ATP5O Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99560DL594
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ATP5O Polyclonal specifically detects ATP5O in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| ATP5O | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, human ATP synthase OSCP subunit, oligomycin sensitivity conferring protein10ATPOATP synthase subunit O, mitochondrial, Oligomycin sensitivity conferral protein, oligomycin sensitivity conferring protein, OSCPhuman ATP synthase OSCP subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ATP5O. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 539 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion