missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ ATP1A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148210
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Human and rat predicted to react based on 100% sequence homology.
The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP-dependent transport of sodium and potassium across the cell membrane. This membrane-bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium/potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families. The different isoforms of the sodium/potassium ATPase exhibit tissue-specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell-specific expression. The alpha-3 subunit has been found in neuronal and to skeletal and cardiac muscle, lung and stomach tissues.
Tekniske data
| ATP1A3 | |
| Polyclonal | |
| Unconjugated | |
| ATP1A3 | |
| AHC2; ATP1A1; Atp1a3; ATPa1a3; Atpa-2; ATPase Na+/K+ transporting subunit alpha 3; ATPase, Na+/K+ transporting, alpha 3 polypeptide; ATPase, Na+K+ transporting, alpha 3 subunit; CAPOS; DYT12; EC 3.6.3.9; Na(+)/K(+) ATPase alpha(III) subunit; Na(+)/K(+) ATPase alpha-3 subunit; Na+, K+ activated adenosine triphosphatase alpha subunit; Na+/K+ ATPase 3; Na+/K+ -ATPase alpha 3 subunit; RDP; Sodium Potassium ATPase; Sodium pump 3; sodium pump subunit alpha-3; sodium/potassium-transporting ATPase alpha-3 chain; sodium/potassium-transporting ATPase subunit alpha-3; sodium-potassium ATPase catalytic subunit alpha-3; sodium-potassium-ATPase, alpha 3 polypeptide | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 232975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q6PIC6 | |
| ATP1A3 | |
| A synthetic peptide corresponding to the N-terminus of the Mouse ATP1A3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion