Få mere at vide
Atorvastatin Lactam Sodium Salt Impurity, TRC
Artikelnummer. 17952355
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17952355 1 mg
17951295 10 mg
17990585 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17952355 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7917751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148217-40-7
Molekylær formel C33 H34 F N2 O6 . Na
Molekylvægt (g/mol) 596.62
Synonym 1H-Pyrrole-1-heptanoic acid, 5-(4-fluorophenyl)-2,3-dihydro-β,δ-dihydroxy-3-(1-methylethyl)-2-oxo-4-phenyl-3-[(phenylamino)carbonyl]-, sodium salt (1:1), (βR,δR,3R)-, 1H-Pyrrole-1-heptanoic acid, 5-(4-fluorophenyl)-2,3-dihydro-β,δ-dihydroxy-3-(1-methylethyl)-2-oxo-4-phenyl-3-[(phenylamino)carbonyl]-, monosodium salt, [3R-[1(βR*,δR*),3R*]]- (9CI)
IUPAC navn sodium;(3R,5R)-7-[(3R)-5-(4-fluorophenyl)-2-oxo-4-phenyl-3-(phenylcarbamoyl)-3-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMIL [Na+].CC(C)[C@@]1(C(=O)Nc2ccccc2)C(=O)N(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])C(=C1c3ccccc3)c4ccc(F)cc4
Tekniske data
Kemisk navn eller materiale Atorvastatin Lactam Sodium Salt Impurity
Analyt- eller komponentnavne Atorvastatin Lactam Sodium Salt Impurity
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C33H35FN2O6.Na/c1-21(2)33(31(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)30(23-13-15-24(34)16-14-23)36(32(33)42)18-17-26(37)19-27(38)20-28(39)40;/h3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;+1/p-1/t26-,27-,33-;/m1./s1
Løsningstype Neat
Industry Type Forensics
Mængde 1 mg
Formel vægt 596.2299
Procent renhed >95%