Få mere at vide
Atorvastatin Di-acetonide tert-Butyl Ester, TRC
Artikelnummer. 30362268
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362268 50 mg
30361709 10 mg
30363077 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362268 Leverandør Toronto Research Chemicals Leverandørnr. TRCD30348150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1116118-82-1
Molekylær formel C50H64FN3O8
Molekylvægt (g/mol) 854.06
Synonym 1,1-Dimethylethyl (4R,6R)-6-[2-[[2-[(4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetyl]amino]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetate
SMIL CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5C[C@H](CC(=O)NCC[C@@H]6C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O6)OC(C)(C)O5
Tekniske data
Kemisk navn eller materiale Atorvastatin Di-acetonide tert-Butyl Ester
Mængde 50 mg
InChI formel InChI=1S/C50H64FN3O8/c1-32(2)45-44(47(57)53-36-18-14-11-15-19-36)43(33-16-12-10-13-17-33)46(34-20-22-35(51)23-21-34)54(45)27-25-38-29-39(60-50(8,9)59-38)30-41(55)52-26-24-37-28-40(61-49(6,7)58-37)31-42(56)62-48(3,4)5/h10-23,32,37-40H,24-31H2,1-9H3,(H,52,55)(H,53,57)/t37-,38-,39-,40-/m1/s1
Formel vægt 853.4677