Få mere at vide
Atorvastatin-d5 Calcium Salt, TRC
Artikelnummer. 30347128
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30347128 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347128 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7917540.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Atorvastatin 3-phenyl-d5 calcium salt
CAS 222412-82-0
Renhedsgrad noter HPLC
Molekylær formel 2 C33 2H5 H29 F N2 O5 . Ca
Mængde 0.5 mg
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-(phenyl-d5)-4-[(phenylamino)carbonyl]-, calcium salt (2:1), (βR,δR)- (9CI), Atorvastatin-d5 calcium salt, (3R,5R)-7-[2-(4-Fluorophenyl)-5-isopropyl-3-(phenyl-d5)-4-phenylcarbamoylpyrrol-1-yl]-3,5-dihydroxyheptanoic acid calcium salt, (βR,δR)-2-(p-Fluorophenyl)-β,δ-dihydroxy-5-isopropyl-3-(phenyl-d5)-4-(phenylcarbamoyl)pyrrole-1-heptanoic acid calcium salt, Atorvastatin-D5 calcium salt (3-phenyl-D5)
InChI formel InChI=1S/2C33H35FN2O5.Ca/c2*1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;/h2*3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;;+2/p-2/t2*26-,27-;/m11./s1/i2*3D,5D,6D,9D,10D;
SMIL [Ca+2].[2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c(C(=O)Nc3ccccc3)c(C(C)C)n(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c2c4ccc(F)cc4.[2H]c5c([2H])c([2H])c(c([2H])c5[2H])c6c(C(=O)Nc7ccccc7)c(C(C)C)n(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c6c8ccc(F)cc8
IUPAC navn calcium;(3R,5R)-7-[2-(4-fluorophenyl)-3-(2,3,4,5,6-pentadeuteriophenyl)-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
Molekylvægt (g/mol) 1165.4
Formel vægt 1164.5157
Procent renhed >95