Få mere at vide
Atomoxetine-d3 Hydrochloride, TRC
Artikelnummer. 30342616
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30342616 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342616 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7914021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Atomoxetine-d3 Hydrochloride
CAS 1217776-38-9
Molekylær formel C17H19D3ClNO
Mængde 1 mg
Synonym (R)-N-(Methyl-d3)-(2-methylphenoxy)benzenepropanamine Hydrochloride, R-Tomexetine-d3 Hydrochloride, Tomoxetine-d3 Hydrochloride, LY 139603-d3, Strattera-d3
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1/i2D3;
SMIL CC1=C(O[C@@H](C2=CC=CC=C2)CCNC([2H])([2H])[2H])C=CC=C1.Cl
IUPAC navn (3R)-3-(2-methylphenoxy)-3-phenyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 294.83