missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ATG12 Rabbit anti-Human, Mouse, Rat, Clone: 9R8W2, Novus Biologicals™
Rabbit Monoclonal Antibody
1450.00 DKK - 3780.00 DKK
Tekniske data
| Antigen | ATG12 |
|---|---|
| Klon | 9R8W2 |
| Fortynding | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Applikationer | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18310502
|
Novus Biologicals
NBP3-15812-20UL |
20 μg |
1450.00 DKK
20µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18341815
|
Novus Biologicals
NBP3-15812-100UL |
100 μg |
3780.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
ATG12 Monoclonal antibody specifically detects ATG12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Tekniske data
| ATG12 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 61-140 of human ATG12 (O94817). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9R8W2 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 9140 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel