missing translation for 'onlineSavingsMsg'
Få mere at vide

Atazanavir-d6, TRC

Product Code. 30329126
Change view
Click to view available options
Mængde:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30329126 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30329126 Supplier Toronto Research Chemicals Supplier No. TRCA7900562.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Atazanavir-d6
CAS 1092540-50-5
Molekylær formel C38H46D6N6O7
Mængde 2.5 mg
Synonym (3S,8S,9S,12S)-,12-Bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-Pentaazatetradecanedioic Acid 14- di(methyl-d3) Ester
InChI formel InChI=1S/C38H52N6O7/c1-37(2,3)31(41-35(48)50-7)33(46)40-29(22-25-14-10-9-11-15-25)30(45)24-44(43-34(47)32(38(4,5)6)42-36(49)51-8)23-26-17-19-27(20-18-26)28-16-12-13-21-39-28/h9-21,29-32,45H,22-24H2,1-8H3,(H,40,46)(H,41,48)(H,42,49)(H,43,47)/t29-,30-,31+,32+/m0/s1/i7D3,8D3
SMIL O=C(NC(C(C)(C)C)C(NC(CC1=CC=CC=C1)[C@@H](O)CN(CC2=CC=C(C3=NC=CC=C3)C=C2)NC([C@H](C(C)(C)C)NC(OC([2H])([2H])[2H])=O)=O)=O)OC([2H])([2H])[2H]
IUPAC navn trideuteriomethyl N-[(2S)-1-[2-[(2S,3S)-3-[[(2S)-3,3-dimethyl-2-(trideuteriomethoxycarbonylamino)butanoyl]amino]-2-hydroxy-4-phenylbutyl]-2-[(4-pyridin-2-ylphenyl)methyl]hydrazinyl]-3,3-dimethyl-1-oxobutan-2-yl]carbamate
Molekylvægt (g/mol) 710.89
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.