Learn More
ASTM Method D5769 Quality Control Reference Material with 3 IS in Isooctane, Dr. Ehrenstorfer
Product Code. 30309665
Change view
Click to view available options
Quantity:
10 x 2 mL
Unit Size:
20mL
Product Code. Quantity Unit Size
30309665 10 x 2 mL 20mL
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309665 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000132IO
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Benzene-D6∣Ethylbenzene∣Ethylbenzene-d10∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣Naphthalene D8∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene
CAS 95-93-2, 95-63-6, 540-84-1, 71-43-2, 1076-43-3, 100-41-4
Analyte or Component Names 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Benzene-D6∣Ethylbenzene∣Ethylbenzene-d10∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣Naphthalene D8∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 10 x 2 mL
Concentration or Composition (by Analyte or Components) Naphthalene D8 1 wt%∣m-Xylene (1,3-Dimethylbenzene) 3 wt%∣o-Xylene (1,2-Dimethylbenzene) 3 wt%∣n-Dodecane 5 wt%∣2,2,4-Trimethylpentane 11.9 wt%∣n-Decane 12 wt%∣n-Hexane 12 wt%∣n-Heptane 16.9 wt%∣n-Octane 16.9 wt%∣Benzene 10000 mg/kg∣Naphthalene 10000 mg/kg∣1,2,4,5-Tetramethylbenzene 20000 mg/kg∣Benzene-D6 20000 mg/kg∣Ethylbenzene-d10 20000 mg/kg∣1,2,4-Trimethylbenzene 30000 mg/kg∣Ethylbenzene 30000 mg/kg∣Toluene 90000 mg/kg