Learn More
Hecht Karl™ Assistent™ Borosilicate Weighing Bottles
Click to view available options
Capacity (Metric):
30 mL
45 mL
Specifications
Closure Type | NS Cover |
Product Type | Weighing Bottle |
Capacity (Metric) | 30 mL |
Includes | Interchangeable NS cover and frosted label |
Material | Borosilicate Glass |