Få mere at vide
Aspartic Acid Diethyl Ester, TRC
Artikelnummer. 30179549
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179549 25 g
30179525 10 g
30179252 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179549 Leverandør Toronto Research Chemicals Leverandørnr. TRCA79017025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 43101-48-0
Molekylær formel C8 H15 N O4 . HCl
Molekylvægt (g/mol) 225.62
Synonym DL-Aspartic Acid Diethyl Ester, Diethyl 2-Aminosuccinate, Diethyl DL-Aspartate;
IUPAC navn diethyl aspartate hydrochloride
SMIL CCOC(CC(C(OCC)=O)N)=O.Cl
Tekniske data
Kemisk navn eller materiale Aspartic Acid Diethyl Ester
InChI formel InChI=1S/C8H15NO4.ClH/c1-3-12-7(10)5-6(9)8(11)13-4-2;/h6H,3-5,9H2,1-2H3;1H
Mængde 25 g
Formel vægt 225.08