Få mere at vide
Aspartame-d5, TRC
Artikelnummer. 30342618
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30342618 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342618 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7900181MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aspartame-d5
CAS 1356849-17-6
Renhedsgrad noter HPLC
Molekylær formel C14H13D5N2O5
Mængde 1 mg
Synonym Aspartame D5 (Phenyl D5)
InChI formel InChI=1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C[C@H](NC(=O)[C@@H](N)CC(=O)O)C(=O)OC)c([2H])c1[2H]
IUPAC navn (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-(2,3,4,5,6-pentadeuteriophenyl)propan-2-yl]amino]-4-oxobutanoic acid
Molekylvægt (g/mol) 299.33
Procent renhed >95