Få mere at vide
Aspartame-D3, TRC
Artikelnummer. 30343651
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30343651 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343651 Leverandør Toronto Research Chemicals Leverandørnr. TRCA79001710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aspartame-d3
CAS 1356841-28-5
Molekylær formel C14H15D3N2O5
Mængde 10 mg
InChI formel InChI=1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])OC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(=O)O
IUPAC navn (3S)-3-amino-4-oxo-4-[[(2S)-1-oxo-3-phenyl-1-(trideuteriomethoxy)propan-2-yl]amino]butanoic acid
Molekylvægt (g/mol) 297.32