missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66909.|D

Additional Details : CAS Number : 98849-88-8 Weight : 0.00001kg

 View more versions of this product

Product Code. 15488459

  • 313.23 DKK / 0.50mg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

1013.00
0.5mg
DYKDDDDK Peptide
-30° to -10°C
Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
HPLC
Lyophilized powder
RUO
Flag PeptideÖ Sigma-Aldrich
Lyophilized powder
Videos
SDS Documents

For Research Use Only