Få mere at vide
Asomate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Asomate |
| CAS | 3586-60-5 |
| Molekylær formel | C9 H18 As N3 S6 |
| Mængde | 100 mg |
| Synonym | Carbamic acid, dimethyldithio-, tris(anhydrosulfide) with (HS)3As (7CI), Carbamic acid, dimethyldithio-, tris(anhydrosulfide) with thioarsenious acid (6CI), Carbamic acid, dimethyldithio-, tris(anhydrosulfide) with thioarsenious acid (H3AsS3) (8CI), Carbamodithioic acid, dimethyl-, tris(anhydrosulfide) with arsenotrithious acid (9CI), Thioarsenious acid (H3AsS3), tris(anhydrosulfide) with dimethyldithiocarbamic acid (8CI), Asomate, TDDA, Tris(dimethyldithiocarbamato)arsenic, Tris(dimethyldithiocarbamoyl)arsine, TTCA |
| InChI formel | InChI=1S/C9H18AsN3S6/c1-11(2)7(14)17-10(18-8(15)12(3)4)19-9(16)13(5)6/h1-6H3 |
| SMIL | CN(C)C(=S)S[As](SC(=S)N(C)C)SC(=S)N(C)C |
| IUPAC navn | bis(dimethylcarbamothioylsulfanyl)arsanyl N,N-dimethylcarbamodithioate |
| Molekylvægt (g/mol) | 435.57 |
| Formel vægt | 434.9041 |