Få mere at vide
Asenapine-13C,d3, TRC
Artikelnummer. 30333298
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30333298 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333298 Leverandør Toronto Research Chemicals Leverandørnr. TRCA78800210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Asenapine-methyl-13C,d3
CAS 1217729-73-1
Renhedsgrad noter HPLC
Molekylær formel C1613CH13D3ClNO
Mængde 10 mg
Synonym 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-2-(methyl-13C-d3)-, (3aR,12bR)-rel- (ACI), rel-(3aR,12bR)-5-Chloro-2,3,3a,12b-tetrahydro-2-(methyl-13C-d3)-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole (ACI), Asenapine-13C,d3, Asenapine-13C,D3 (N-methyl-13C,D3)
InChI formel InChI=1S/C17H16ClNO/c1-19-9-14-12-4-2-3-5-16(12)20-17-7-6-11(18)8-13(17)15(14)10-19/h2-8,14-15H,9-10H2,1H3/t14-,15-/m0/s1/i1+1D3
SMIL [2H][13C]([2H])([2H])N1C[C@@H]2[C@@H](C1)c3cc(Cl)ccc3Oc4ccccc24
IUPAC navn (2R,6R)-9-chloro-4-(trideuterio(113C)methyl)-13-oxa-4-azatetracyclo[12.4.0.02,6.07,12]octadeca-1(18),7(12),8,10,14,16-hexaene
Molekylvægt (g/mol) 289.78
Procent renhed >95