Få mere at vide
ASC 67131, TRC
Artikelnummer. 30179417
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179417 1 g
30179154 500 mg
30179089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179417 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1012601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 697298-57-0
Molekylær formel C9 H22 N O4 P S2
Molekylvægt (g/mol) 303.38
Synonym Phosphoramidothioic acid, [2-(methylsulfonyl)ethyl]-, O-ethyl S-(1-methylpropyl) ester (9CI), ASC 67131
IUPAC navn N-[butan-2-ylsulfanyl(ethoxy)phosphoryl]-2-methylsulfonylethanamine
SMIL CCOP(=O)(NCCS(=O)(=O)C)SC(C)CC
Tekniske data
Kemisk navn eller materiale Fosthiazate metabolite 1 ASC-67131
Analyt- eller komponentnavne Fosthiazate metabolite 1 ASC-67131
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C9H22NO4PS2/c1-5-9(3)16-15(11,14-6-2)10-7-8-17(4,12)13/h9H,5-8H2,1-4H3,(H,10,11)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 g
Formel vægt 303.0728