Få mere at vide
Tocris Bioscience™ AS2717638
Beskrivelse
AS2717638 is a potent LPA5 receptor antagonist (IC50 = 38 nM in BV-2 microglia cells). It is selective for LPA5 over LPA1, LPA2, and LPA3. It inhibits LPA-mediated pro-inflammatory transcription factor phosphorylation. In vivo, AS2717638 inhibits LPA5 agonist-induced allodynia in mice. It also exhibits analgesic effects against both neuropathic and inflammatory pain in rodent models. Orally bioavailable and brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 6,7-Dimethoxy-2-(5-methyl-1,2-benzisoxazol-3-yl)-4-(1-piperidinylcarbonyl)-1(2 H)-isoquinolinone |
| CAS | 2148339-28-8 |
| Mængde | 10 mg |
| Mål | LPAR5/LPA5/GPR92 |
| Molekylær formel | C25H25N3O5 |
| Renhed | 0.98 |