missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ AS 99
GREENER_CHOICE

Artikelnummer. 18765955
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18765955 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18765955 Leverandør Tocris Bioscience™ Leverandørnr. 7648/5

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Selective ASH1L histone methyltransferase inhibitor

AS 99 is a selective ASH1L histone methyltransferase inhibitor (IC50 = 0.79 μM); it binds to the autoinhibitory loop region in the SET domain (Kd = 0.89 μM). AS 99 exhibits >100-fold selectivity for ASH1L over a range of lysine methyltransferases; no significant inhibition is observed at 50 μM against a panel of 20 histone methyltransferases, including NSD1, NSD2, NSD3, and SETD2. In leukemia cells, AS 99 blocks H3K36 methylation, and causes a dose-dependent downregulation of MLL fusion target genes; it also induces apoptosis and differentiation and inhibits cell proliferation. AS 99 reduces leukemia burden in the xenotransplantation mouse model of MLL leukemia without affecting blood count in normal mice.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-[[3-[3-(Aminothioxomethyl)phenyl]-1-[1-[(trifluoromethyl)sulfonyl]-4-piperidinyl]-1 H-indol-6-yl]methyl]-1-methyl-3-azetidinecarboxamide
CAS 2323623-93-2
Mængde 5 mg
Mål Lysine Methyltransferase Inhibitors
Molekylær formel C27H30F3N5O3S2
Renhed 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.