missing translation for 'onlineSavingsMsg'
Få mere at vide

Articaine hydrochloride, MedChemExpress

Artikelnummer. 16452166
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16452166 500 mg
16408356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16452166 Leverandør Medchem Express Leverandørnr. HYB0516

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Articaine hydrochloride (Hoe-045) is an amide anaesthetic containing an ester group, reversibly binding to the α-subunit of the voltage-gated sodium channels within the inner cavity of the nerve, can provide effective pain relief. Articaine hydrochloride ameliorates LPS-induced acute kidney injury via inhibition of NF-ĸB activation and the NLRP3 inflammasome pathway.

Articaine hydrochloride (Hoe-045) is an amide anaesthetic containing an ester group, reversibly binding to the α-subunit of the voltage-gated sodium channels within the inner cavity of the nerve, can provide effective pain relief. Articaine hydrochloride ameliorates LPS-induced acute kidney injury via inhibition of NF-ĸB activation and the NLRP3 inflammasome pathway.

Tekniske data

Kemisk navn eller materiale Articaine hydrochloride
CAS 23964-57-0
Farve White
Renhedsgrad noter Research
Molekylær formel C13H21ClN2O3S
Mængde 500 mg
Synonym Hoe-045
Opløselighedsinformation H2O : 100 mg/mL (311.68 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (311.68 mM; Need ultrasonic)
SMIL O=C(C(C)NCCC)NC(C(C)=CS1)=C1C(OC)=O.Cl
Molekylvægt (g/mol) 320.84
Formel vægt 320.84
Procent renhed 98.46%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.