Få mere at vide
Articaine Hydrochloride, LoGiCal
Artikelnummer. 30462337
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462337 10 mg
1 options
Artikelnummer. 30462337 Leverandør LoGiCal Leverandørnr. LGCFOR0369.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Articaine hydrochloride
CAS 23964-57-0
Analyt- eller komponentnavne Articaine hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O3 S . Cl H
Certificeringer/compliance ISO 9001
Synonym Articaine Hydrochloride, 2-Thiophenecarboxylic acid, 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-, methyl ester, hydrochloride (1:1), 2-Thiophenecarboxylic acid, 4-methyl-3-[2-(propylamino)propionamido]-, methyl ester, monohydrochloride (8CI), 2-Thiophenecarboxylic acid, 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-, methyl ester, monohydrochloride (9CI), Carticain, Carticaine chlorhydrate, Carticaine hydrochloride, HOE 045, HOE 40045, Methyl 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylate hydrochloride, Septanest, Ubistesine, Ultacain, Ultracain, Ultracaine
InChI formel InChI=1S/C13H20N2O3S.ClH/c1-5-6-14-9(3)12(16)15-10-8(2)7-19-11(10)13(17)18-4;/h7,9,14H,5-6H2,1-4H3,(H,15,16);1H
Løsningstype Neat
SMIL Cl.CCCNC(C)C(=O)Nc1c(C)csc1C(=O)OC
IUPAC navn methyl 4-methyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxylate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 320.84
Mængde 10 mg
Formel vægt 320.0961 g/mol