Få mere at vide
Articaine EP Impurity E (as Hydrochloride), Mikromol™
Beskrivelse
Articaine EP Impurity E (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isopropylarticaine Hydrochloride |
| CAS | 2983831-71-4 |
| Analyt- eller komponentnavne | Isopropylarticaine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H20 N2 O3 S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate Hydrochloride (Isopropylarticaine Hydrochloride), Articaine Hydrochloride Imp. E (EP), Articaine Imp. E (EP), Articaine USP Related Compound E, Articaine USP RC E, Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. E (EP) as Hydrochloride, Isopropylarticaine Hydrochloride, Articaine Related Compound E as Hydrochloride, Articaine Hydrochloride Impurity E as Hydrochloride, Articaine Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C23H32ClN3.2ClH/c1-25(2)14-6-15-26(3)16-7-17-27-22-9-5-4-8-19(22)10-11-20-12-13-21(24)18-23(20)27;;/h4-5,8-9,12-13,18H,6-7,10-11,14-17H2,1-3H3;2*1H |
| SMIL | Cl.COC(=O)c1scc(C)c1NC(=O)C(C)NC(C)C |
| IUPAC navn | methyl 4-methyl-3-[2-(propan-2-ylamino)propanoylamino]thiophene-2-carboxylate;hydrochloride |
| Vis mere |