Få mere at vide
Articaine EP Impurity A (as Hydrochloride), Mikromol™
Artikelnummer. 17931566
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17931566 25 mg
17962986 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17931566 Leverandør Mikromol Leverandørnr. MM0369.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Articaine EP Impurity A (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetamidoarticaine Hydrochloride
CAS 2984442-47-7
Analyt- eller komponentnavne Acetamidoarticaine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C12 H18 N2 O3 S . HCl
Certificeringer/compliance ISO 9001
Synonym Methyl 3-[[2-(Propylamino)acetyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Acetamidoarticaine Hydrochloride), Articaine Hydrochloride Imp. A (EP), Articaine Imp. A (EP), Articaine USP Related Compound A, Articaine USP RC A, Methyl 3-[[2-(Propylamino)acetyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. A (EP) as Hydrochloride
InChI formel InChI=1S/C12H18N2O3S.ClH/c1-4-5-13-6-9(15)14-10-8(2)7-18-11(10)12(16)17-3;/h7,13H,4-6H2,1-3H3,(H,14,15);1H
SMIL Cl.CCCNCC(=O)Nc1c(C)csc1C(=O)OC
IUPAC navn methyl 4-methyl-3-[[2-(propylamino)acetyl]amino]thiophene-2-carboxylate;hydrochloride
Molekylvægt (g/mol) 306.81
Mængde 25 mg
Formel vægt 306.0805 g/mol