Få mere at vide
Artesunate-d4, TRC
Artikelnummer. 30337661
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30337661 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337661 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7778035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Artesunate-d4
CAS 1316753-15-7
Molekylær formel C19H24D4O8
Mængde 5 mg
Synonym Butanedioic Acid Mono(3R,5aS,6R,8aS,9R,10R,12R,12aR)-decahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-yl] Ester-d4, Artesunic Acid-d4
InChI formel InChI=1S/C19H28O8/c1-10-4-5-13-11(2)16(23-15(22)7-6-14(20)21)24-17-19(13)12(10)8-9-18(3,25-17)26-27-19/h10-13,16-17H,4-9H2,1-3H3,(H,20,21)/t10-,11-,12+,13+,16-,17-,18-,19-/m1/s1/i6D2,7D2
SMIL C[C@H]1[C@@](CC[C@@](C)(OO2)O3)([H])[C@]2([C@]3([H])O[C@](OC(C([2H])([2H])C([2H])([2H])C(O)=O)=O)([H])[C@@]4(C)[H])[C@@]4([H])CC1
IUPAC navn 2,2,3,3-tetradeuterio-4-oxo-4-[[(1R,4S,5R,8S,9R,10S,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.04,13.08,13]hexadecan-10-yl]oxy]butanoic acid
Molekylvægt (g/mol) 388.45