missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART3 Antibody (063), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90224AF405
Additional Details : Weight : 0.00970kg
Description
ART3 Monoclonal specifically detects ART3 in Human samples. It is validated for ELISA.Specifications
ART3 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
419 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
63 | |
ELISA | |
ADP-ribosyltransferase 3, EC 2.4.2.31, ecto-ADP-ribosyltransferase 3, FLJ26404, Mono(ADP-ribosyl)transferase 3, mono-ADP-ribosyltransferase, NAD(P)(+)--arginine ADP-ribosyltransferase 3, TMART | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ART3 (AAH08397.1; Met1-Ser362). | |
0.1 mL | |
Primary | |
Human | |
Purified |