Få mere at vide
Arsenic (V) Standard: As+5 @ 100 μg/mL in H2O, VHG™
Artikelnummer. 30003092
Skift visning
Mængde:
50 ml
100 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003092 50 ml
17969374 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003092 Leverandør VHG Labs Leverandørnr. VHGSPAS5W50
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Arsenic (V) Standard: As+5 @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Arsenic+5
CAS 17428-41-0
Metoder Speciation
Analyt- eller komponentnavne Arsenic+5
Forsendelsestilstand Room Temperature
Molekylær formel As5+
Synonym As5+
SMIL [As5+]
IUPAC navn Arsenic+5
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 74.92
Mængde 50 ml
Formel vægt 74.92 g/mol