missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
430.00 DKK - 776.00 DKK
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12957323
|
Fisher Chemical
S 603 050 |
50 mL | Bottle |
776.00 DKK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12967323
![]() |
Fisher Chemical
S 603 100 |
100 mL | Bottle |
430.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
7697-37-2 | |
10 mg/L | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
63.01 | |
CHEBI:48107 |
HNO3 | |
MFCD00011349 | |
O[N+]([O-])=O | |
HNO3 | |
944 | |
Arsenic |
Safety and Handling
missing translation for 'shelfLife' : 12 months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Arsenic solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™