Få mere at vide
Arsenic Oxide (As2O5) Hydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Arsenic Oxide (As2O5) Hydrate |
| CAS | 2179255-37-7 |
| Molekylær formel | As2O5 xH2O |
| Mængde | 5 g |
| Synonym | Arsenic (V) oxide Hydrate, Arsenic anhydride Hydrate, Arsenic oxide (As4O10) Hydrate, Arsenic pentoxide Hydrate, Arsenic(V) pentoxide Hydrate, Diarsenic pentoxide Hydrate, Diarsonic pentoxide Hydrate |
| InChI formel | InChI=1S/As2O5/c3-1(4)2(5,6)7 |
| SMIL | [O][As](=O)([O])[As](=O)=O |
| IUPAC navn | arsoarsonic acid |
| Molekylvægt (g/mol) | 231.86 |
| Formel vægt | 229.8178 |