Få mere at vide
Arsenic AAS Standard, 1000 mg/L As in HNO3 2%, Honeywell Fluka™
Artikelnummer. 16654264 Shop alle Honeywell Fluka produkter
Skift visning
Mængde:
100 ml
500 ml
Pakningsstørrelse:
500ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16654264 500 ml 500ml
16644264 100 ml -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 16654264 Leverandør Honeywell Fluka Leverandørnr. E4004500ML
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Kemisk navn eller materiale Arsenic AAS Standard
Tæthed 1,015 g/cm3
pH <2
Mængde 500 ml