Få mere at vide
Novus Biologicals™ Aromatase Overexpression Lysate
Artikelnummer. 18099669 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Pakningsstørrelse:
0.1mg
Artikelnummer. Mængde Pakningsstørrelse
18099669 0.1 mg 0.1mg
1 options
Artikelnummer. 18099669 Leverandør Novus Biologicals™ Leverandørnr. NBP211585
 Begrænset vare
Forventet forsendelse: 15-10-2026
The Aromatase Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_11253. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens Aromatase, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only