Få mere at vide
Aroclor 1016, SPEX CertiPrep™
Artikelnummer. 12919344 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12919344 1 mL 1ml
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 12919344 Leverandør SPEX Certiprep Leverandørnr. S310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Aroclor 101612674-11-21000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Aroclor 1016
Emballage 2mL amber prescored glass ampule
CAS 12674-11-2, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C12H7Cl3
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle RIBGNAJQTOXRDK-UHFFFAOYSA-N
SMIL C1=CC(=CC(=C1)Cl)C2=CC(=CC(=C2)Cl)Cl
IUPAC navn 1,3-dichloro-5-(3-chlorophenyl)benzene
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 257.538
PubChem CID 38037
Mængde 1 mL
Vis mere Vis mindre
Product intended for laboratory use only