Få mere at vide
Novus Biologicals™ ARMC8 Overexpression Lysate
Artikelnummer. 18271465 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18271465 0.1 mg
1 options
Artikelnummer. 18271465 Leverandør Novus Biologicals™ Leverandørnr. NBL107710
 Begrænset vare
Forventet forsendelse: 09-10-2026
The ARMC8 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC215221 and based on accession number NM_213654. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens armadillo repeat containing 8 (ARMC8), transcript variant 3, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only