missing translation for 'onlineSavingsMsg'
Få mere at vide

Aripiprazole Hydrate, TRC

Artikelnummer. 30340696
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340696 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30340696 Leverandør Toronto Research Chemicals Leverandørnr. TRCA771005250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Aripiprazole Hydrate
CAS 503445-20-3
Molekylær formel C23 H27 Cl2 N3 O2. x[H2O]
Mængde 250 mg
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone Hydrate, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydrocarbostyril Hydrate, Abilify Hydrate, Abilitat Hydrate, OPC 14597 Hydrate, OPC 31 Hydrate, 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-, hydrate, Aripiprazole hydrate
InChI formel InChI=1S/C23H27Cl2N3O2.H2O/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-6,8,16H,1-2,7,9-15H2,(H,26,29);1H2
SMIL O=C1NC2=CC(OCCCCN3CCN(CC3)C4=C(Cl)C(Cl)=CC=C4)=CC=C2CC1.O
IUPAC navn 7-(4-(4-(2,3-dichlorophenyl)piperazin-1-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one hydrate
Molekylvægt (g/mol) 466.4
Formel vægt 465.16
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.