missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP28 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99176H
Additional Details : Weight : 0.00970kg
Description
ARHGAP28 Polyclonal specifically detects ARHGAP28 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGAP28 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
79822 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
DKFZp686A2038, FLJ10312, KIAA1314FLJ27160, Rho GTPase activating protein 28, rho GTPase-activating protein 28, Rho-type GTPase-activating protein 28 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ARHGAP28. | |
0.1 mL | |
Primary | |
Human | |
Purified |