missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argonaute 4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05866AF647
Additional Details : Weight : 0.00970kg
Description
Argonaute 4 Polyclonal specifically detects Argonaute 4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Argonaute 4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AGO4argonaute 4, Argonaute4, eIF2C 4, eIF-2C 4, Eukaryotic translation initiation factor 2C 4, eukaryotic translation initiation factor 2C, 4, FLJ20033, hAgo4, KIAA1567argonaute4, protein argonaute-4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Argonaute 4. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
192670 | |
Store at 4°C in the dark. | |
IgG |