Få mere at vide
Arginino-succinic Acid Disodium Salt (>80%), TRC
Artikelnummer. 30134062
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134062 25 mg
30009135 5 mg
30011876 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134062 Leverandør Toronto Research Chemicals Leverandørnr. TRCA76920025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 918149-29-8
Molekylær formel C10H16N4Na2O6
Molekylvægt (g/mol) 334.24
Synonym N-[[[(4S)-4-Amino-4-carboxybutyl]amino]iminomethyl]-L-aspartic Acid Disodium Salt
IUPAC navn sodium Nw-((S)-2-carboxy-1-carboxylatoethyl)-L-argininate
SMIL N=C(NCCC[C@H](N)C(O[Na])=O)N[C@H](C(O[Na])=O)CC(O)=O
Tekniske data
Kemisk navn eller materiale Arginino-succinic Acid Disodium Salt
Analyt- eller komponentnavne Arginino-succinic Acid Disodium Salt
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C10H18N4O6.2Na/c11-5(8(17)18)2-1-3-13-10(12)14-6(9(19)20)4-7(15)16;;/h5-6H,1-4,11H2,(H,15,16)(H,17,18)(H,19,20)(H3,12,13,14);;/q;2*+1/p-2
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 25 mg
Formel vægt 334.09