Få mere at vide
Arginase inhibitor 1, MedChemExpress
Artikelnummer. 16449367
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16449367 5 mg
16422897 50 mg
16406936 100 mg
16406828 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16449367 Leverandør Medchem Express Leverandørnr. HY157755mg
MedChemExpress Arginase inhibitor 1 is a potent inhibitor of human arginases I and II with IC50s of 223 and 509 nM, respectively.
Arginase inhibitor 1 is a potent inhibitor of human arginases I and II with IC50s of 223 and 509 nM, respectively.
Tekniske data
Kemisk navn eller materiale Arginase inhibitor 1
Fysisk form Solid
CAS 1345808-25-4
Farve Off-White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C13H27BN2O4
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : ≥ 48 mg/mL (167.73 mM) ∣H2O : ≥ 30 mg/mL (104.83 mM)
SMIL O=C(O)[C@@](CCN1CCCCC1)(N)CCCCB(O)O
Molekylvægt (g/mol) 286.18
Formel vægt 286.18
Procent renhed 98.0%