Få mere at vide
Tocris Bioscience™ Arformoterol Tartrate
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 6219/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Arformoterol tartrate is a long-acting β2 adrenergic agonist (LABA). Bronchodilator.Tekniske data
| Long-acting Agonist (LABA) | |
| 200815-49-2 | |
| 494.49g/mol | |
| Adrenergic beta-2 Receptor Agonists |
| Adrenergic beta-2 | |
| C19H24N2O4.C4H6O6 | |
| Store at 4°C | |
| 10 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only