Få mere at vide
Arbidol Hydrochloride, TRC
Artikelnummer. 30137574
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137574 10 mg
30012753 250 mg
30009350 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137574 Leverandør Toronto Research Chemicals Leverandørnr. TRCA76600010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131707-23-8
Molekylær formel C22 H25 Br N2 O3 S . Cl H
Molekylvægt (g/mol) 513.88
Synonym 1H-Indole-3-carboxylic acid, 6-bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-[(phenylthio)methyl]-, ethyl ester, hydrochloride (1:1), 1H-Indole-3-carboxylic acid, 6-bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-[(phenylthio)methyl]-, ethyl ester, monohydrochloride (9CI), Arbidol, Arbidol hydrochloride
IUPAC navn ethyl 6-bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-(phenylsulfanylmethyl)indole-3-carboxylate;hydrochloride
SMIL Cl.CCOC(=O)c1c(CSc2ccccc2)n(C)c3cc(Br)c(O)c(CN(C)C)c13
Tekniske data
Kemisk navn eller materiale Arbidol hydrochloride
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C22H25BrN2O3S.ClH/c1-5-28-22(27)20-18(13-29-14-9-7-6-8-10-14)25(4)17-11-16(23)21(26)15(19(17)20)12-24(2)3;/h6-11,26H,5,12-13H2,1-4H3;1H
Formel vægt 512.0536
Procent renhed >95%