Learn More
Arachidonic Acid, TRC
Product Code. 30343551
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30343551 100 mg 100mg
1 options
Product Code. 30343551 Supplier Toronto Research Chemicals Supplier No. TRCA765000100MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Arachidonic Acid
CAS 506-32-1
Molecular Formula C20 H32 O2
Quantity 100 mg
Synonym 5,8,11,14-Eicosatetraenoic acid, (all-Z)- (8CI), (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic acid, (all-Z)-5,8,11,14-Eicosatetraenoic acid, 20:4n-6, 5,8,11,14-all-cis-Eicosatetraenoic acid, 5-cis,8-cis,11-cis,14-cis-Eicosatetraenoic acid, 5Z,8Z,11Z,14Z-Eicosatetraenoic acid, A 0871, Arachidonic acid, Immunocytophyt, Immunocytophyte, Vevodar, all-cis-5,8,11,14-Eicosatetraenoic acid, cis-δ5,8,11,14-Eicosatetraenoic acid
InChI Formula InChI=1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)O
IUPAC Name (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
Molecular Weight (g/mol) 304.47
Formula Weight 304.2402