Få mere at vide
Arachidonic Acid-d8 (Major, 10mg/ml in Methyl Acetate), TRC
Artikelnummer. 30340268
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30340268 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340268 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7650045MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Arachidonic Acid-d8
CAS 69254-37-1
Molekylær formel C20H24D8O2
Mængde 5 mg
Synonym (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic-5,6,8,9,11,12,14,15-d8 Acid, (all-Z)-5,8,11,14-Eicosatetraenoic-5,6,8,9,11,12,14,15-d8 Acid, Arachidonic Acid-5,6,8,9,11,12,14,15-2H8
InChI formel InChI=1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-/i6D,7D,9D,10D,12D,13D,15D,16D
SMIL CCCCC/C=C\C/C=C\C/C=C\C/C=C\CCCC(=O)O
IUPAC navn (5Z,8Z,11Z,14Z)-5,6,8,9,11,12,14,15-octadeuterioicosa-5,8,11,14-tetraenoic acid
Molekylvægt (g/mol) 312.52
Koncentration eller sammensætning (efter analyt eller komponenter) 10 mg/mL