Få mere at vide
Tocris Bioscience™ AR-C 118925XX
Selective and competitive P2Y2 antagonist
Brand: Tocris Bioscience™ 4890/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
AR-C 118925XX is a selective, competitive P2Y2 receptor antagonist. Inactive against a panel of 37 other receptors at a concentration of 10 μM. Inhibits ATP-γS-induced mucin secretion in bronchial epithelial cells (IC50 = 1 μM). Inhibits P2Y2 receptor-induced β-arrestin translocation in vitro (pA2 = 37.2-51.3 nM).Tekniske data
| 5-[[5-(2,8-Dimethyl-5H-dibenzo[a,d]cyclohepten-5-yl)-3,4-dihydro-2-oxo-4-thioxo-1(2H)-pyrimidinyl]methyl]-N-2H-tetrazol-5-yl-2-furancarboxamide | |
| 216657-60-2 | |
| 5 mg |