missing translation for 'onlineSavingsMsg'
Få mere at vide

(aR)-a-Methyl-N-[[3-(trifluoromethyl)phenyl]methyl]-1-naphthalenemethanamine Hydrochloride, TRC

Artikelnummer. 30482097
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482097 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30482097 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33095050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (alphaR)-alpha-Methyl-N-[(3-(trifluoromethyl)phenyl)methyl]-1-naphthalenemethanamine Hydrochloride
CAS 1229225-42-6
Molekylær formel C20 H18 F3 N . Cl H
Mængde 50 mg
Synonym 1-Naphthalenemethanamine, α-methyl-N-[[3-(trifluoromethyl)phenyl]methyl]-, hydrochloride (1:1), (αR)-
InChI formel InChI=1S/C20H18F3N.ClH/c1-14(18-11-5-8-16-7-2-3-10-19(16)18)24-13-15-6-4-9-17(12-15)20(21,22)23;/h2-12,14,24H,13H2,1H3;1H/t14-;/m1./s1
SMIL Cl.C[C@@H](NCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
IUPAC navn (1R)-1-naphthalen-1-yl-N-[[3-(trifluoromethyl)phenyl]methyl]ethanamine;hydrochloride
Molekylvægt (g/mol) 365.82
Formel vægt 365.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.