Få mere at vide
AR-9281, MedChemExpress
Beskrivelse
AR9281 is a potent and selective inhibitor of soluble epoxide hydrolase (s-EH), with potential for the treatment of hypertension and type 2 diabetes.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | AR-9281 |
| CAS | 913548-29-5 |
| Farve | Off-White |
| Renhedsgrad noter | Research |
| Molekylær formel | C18H29N3O2 |
| Mængde | 5 mg |
| Synonym | APAU |
| Opløselighedsinformation | DMSO : 25 mg/mL (78.26 mM; Need ultrasonic) |
| SMIL | O=C(NC1(C2)CC3CC2CC(C3)C1)NC4CCN(C(C)=O)CC4 |
| Molekylvægt (g/mol) | 319.44 |
| Vis mere |