missing translation for 'onlineSavingsMsg'
Få mere at vide

AQ-13 dihydrochloride, MedChemExpress

Artikelnummer. 16492958
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16492958 10 mg 10mg
16472556 25 mg -
16415707 5 mg -
16409536 50 mg -
16404286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16492958 Leverandør Medchem Express Leverandørnr. HY100358

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress AQ-13 dihydrochloride is an aminoquinoline antimalarial drug that is effective against drug-resistant strains of Plasmodium falciparum.

AQ-13 dihydrochloride is an aminoquinoline antimalarial drug that is effective against drug-resistant strains of Plasmodium falciparum.

Tekniske data

Kemisk navn eller materiale AQ-13 dihydrochloride
Fysisk form Solid
CAS 169815-40-1
Farve Earth Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C16H24Cl3N3
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation H2O : 100 mg/mL (274.17 mM; Need ultrasonic) ∣DMSO : 80 mg/mL (219.33 mM; Need ultrasonic)
SMIL CCN(CC)CCCNC1=CC=NC2=CC(Cl)=CC=C12.[H]Cl.[H]Cl
Molekylvægt (g/mol) 364.74
Formel vægt 364.74
Procent renhed 98.08%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.