missing translation for 'onlineSavingsMsg'
Learn More

Apremilast-d5, TRC

Product Code. 30342398
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30342398 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30342398 Supplier Toronto Research Chemicals Supplier No. TRCA72970210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Apremilast-d5
CAS 1258597-47-5
Renhedsgrad noter HPLC
Molekylær formel C22 D5 H19 N2 O7 S
Mængde 10 mg
Synonym (S)-2-[1-(3-Ethoxy-d5-4-methoxyphenyl)-2-methylsulfonylethyl]-4-acetylaminoisoindoline-1,3-dione, (S)-N-(2-(1-(3-Ethoxy-d5-4-methoxyphenyl)-2-(methylsulfonyl)ethyl)-1,3-dioxoisoindolin-4-yl)acetamide, Apremilast-d5, CC 10004-d5
InChI formel InChI=1S/C22H24N2O7S/c1-5-31-19-11-14(9-10-18(19)30-3)17(12-32(4,28)29)24-21(26)15-7-6-8-16(23-13(2)25)20(15)22(24)27/h6-11,17H,5,12H2,1-4H3,(H,23,25)/t17-/m1/s1/i1D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1cc(ccc1OC)[C@@H](CS(=O)(=O)C)N2C(=O)c3cccc(NC(=O)C)c3C2=O
IUPAC navn N-[2-[(1S)-1-[4-methoxy-3-(1,1,2,2,2-pentadeuterioethoxy)phenyl]-2-methylsulfonylethyl]-1,3-dioxoisoindol-4-yl]acetamide
Molekylvægt (g/mol) 465.531
Formel vægt 465.162
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.